Learn More
Eisco™ Glass Equilibrium Tubes
SCI_ED
Catalog No. S23740 Shop All Eisco Products
Change view
Click to view available options
L x W x H: 10.2 x 6.7 x 1.6 in.
Catalog No.
S23740 L x W x H: 10.2 x 6.7 x 1.6 in.
1 options
Catalog No. S23740 Supplier Eisco™ Supplier No. PH0202C
Add to Cart
Add to Cart
For demonstration of the phenomena of capillarity and surface tension.
  • With plastic base stand
  • Consists of set of five interconnecting glass tubes of different diameters
  • Includes capillary tube and plastic base
Specifications
Product Type Glass Equilibrium Tubes
For Use With (Application) For demonstration of the phenomena of capillarity and surface tension
Includes Plastic stand and 5 interconnecting glass capillary tubes
Dimensions (L x W x H) 10.2 x 6.7 x 1.6 in.