Learn More
Description
Hydrophilic Glass Fiber without Binder Resin filter
Glass Fiber Filters without Binder Resin:
The filters without binder resin retain their structural integrity without weight loss when heated up to 500 °C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.
Type APFA
•Retains fine particles with good efficiency, even at high flow rates
•Recommended for monitoring wastewater and collecting suspended particles in gases (e.g., fumes from chimneys)
•Use in biochemical applications (e.g., collection of cells and filtration of protein or nucleic acid precipitates)
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | White |
| Filter Type | Depth Filter |
| Material | Glass Fiber |
| Pore Size | 0.7 μm |
| Sterility | Sterile |
| Filter Code | APFF |
| Flow Rate | 1.4 mL/min/ cm2 |
| For Use With (Application) | Use for filtering extremely fine precipitates such as protein, Nucleic Acids, or Serum Precipitates |
| Grade | APFF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.