Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Glass Fiber Filters Without Binder

Catalog No. APFF14250 Shop All MilliporeSigma Products
Change view
Click to view available options
Diameter (Metric):
2.4 cm
2.5 cm
3.7 cm
4.7 cm
7 cm
9 cm
14.2 cm
Grade:
AP40
APFF
Quantity:
100 Pk.
50 Pk.
500 Pk.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Grade Quantity
APFF14250 14.2 cm APFF 50 Pk.
AP4003705 3.7 cm AP40 500 Pk.
AP4002405 2.4 cm AP40 500 Pk.
AP4014250 14.2 cm AP40 50 Pk.
AP4002500 2.5 cm AP40 100 Pk.
AP4004705 4.7 cm AP40 500 Pk.
AP4004700 4.7 cm AP40 100 Pk.
APFF02500 2.5 cm APFF 100 Pk.
APFF09050 9 cm APFF 50 Pk.
AP4009000 9 cm AP40 100 Pk.
AP4007000 7 cm AP40 100 Pk.
APFF04700 4.7 cm APFF 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. APFF14250 Supplier MilliporeSigma™ Supplier No. APFF14250
Add to Cart
Add to Cart

Choose from six grades of filters. Glass Fiber Filters without Binders are made of borosilicate microfiber glass. Available in multiple pore sizes and flow rates for a variety of filtration and analysis applications utilizing depth filters. Autoclavable.

For prefiltration and contamination analysis

OverviewMillipore fibrous disc filters are available in a wide range of flow rates and throughput capacities. Our Glass Fiber filters are available with or without binder resins and can be sterilized by EO, gamma, or autoclave (121 °C at 1 bar).Glass Fiber Filters with Binder Resin:Millipore filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated liquids. They are also widely used for clarification of aqueous solutions.

Type AP15

  • Lowest dirt-holding capacity
  • Recommended prefilter for 0.2 to 0.6 μm filters

Type AP20

  • Lower retention, higher dirt-holding capacity than AP15 filters
  • Recommended prefilter for 0.8 to 8.0 μm filters
  • Use upstream to protect type AP15 filters

Type AP25

  • Added thickness yields same retention and greater dirt-holding capacity as AP20
  • Recommended prefilter for 0.9 to 8 μm filters, especially for proteinaceous and heavily contaminated liquids
  • Use upstream to protect type AP15 filters Glass Fiber Filters without Binder Resin:The filters without binder resin retain their structural integrity without weight loss when heated up to 500°C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.

Type APFA

  • Retains fine particles with good efficiency, even at high flow rates
  • Recommended for monitoring wastewater and collecting suspended particles in gases (e.g., fumes from chimneys)
  • Use in biochemical applications (e.g., collection of cells and filtration of protein or nucleic acid precipitates)

Type APFB

  • Higher mechanical strength when wet and greater loading capacity than type APFA filters
  • Use for liquid clarification, quantification of solids in suspensions of fine particles, and scintillation counting

Type APFC

  • Greater retention, especially for fine particulate and microorganism removal than type APFA filters
  • Use for determining total suspended solids in drinking water
  • Use for filtering proteins or nucleic acid TCA precipitates and for collecting cells and microorganisms

Type APFD

  • Thick filter with high flow and low retention
  • Use for clarifying suspensions containing particulates greater than 1.0 μm

Type APFF

  • Use for filtering extremely fine precipitates such as protein, nucleic acids, or serum precipitates
  • Recommended for USEPA method 1311 for TCLP analysis

Type AP40

  • Recommended for Standard Methods Total Suspended Solids 2540D
  • Recommended for USEPA method 1311 for TCLP analysis
  • Maintains structural integrity without weight loss when ignited to 550°C (1022°F) after sample filtration
  • Recommended for determining volatile suspended matter in wastewater and industrial effluents
TRUSTED_SUSTAINABILITY

Specifications

Airflow Rate 18.6 L/min/ cm2
Air Resistance 120 mm of H2O
Autoclavable Autoclavable
Color White
Filter Type Depth Filter
Material Borosilicate Glass Microfiber
Particle Retention 0.7 μm
Pore Size 0.7 μm
Sterility Sterile
Flow Rate 1.4 mL/min/ cm2
For Use With (Application) Use for filtering extremely fine precipitates such as protein, Nucleic Acids, or Serum Precipitates
Grade APFF
Hydrophobicity Hydrophilic
Quantity 50 Pk.
Surface Type Plain
Temperature (English) Max. 932°F
Temperature (Metric) Max 500°C
Thickness (English) 0.0149 in.
Thickness (Metric) 380 μm
Type Filter Membranes
Wettability Hydrophilic
Diameter (English) 5.59 in.
Diameter (Metric) 14.2 cm
Diameter (Metric) Disc 14.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.