Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Glass-filled Nylon Connectors with PTFE Seals
SDP

Catalog No. p-317103 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size:
13-425 to 13-425
13-425 to 15-425
13-425 to 20-400
15-425 to 20-400
18-400 to 20-400
20-400 to 20-400
20-400 to 22-400
20-400 to 24-400
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size
K7472051820 18-400 to 20-400
K7472051520 15-425 to 20-400
K747205-1313 13-425 to 13-425
K747205-1320 13-425 to 20-400
K7472051315 13-425 to 15-425
K7472052020 20-400 to 20-400
K7472052022 20-400 to 22-400
K7472052024 20-400 to 24-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. K7472051820 Supplier DWK Life Sciences Supplier No. 7472051820
Add to Cart
Add to Cart

Kimble™ Glass-filled Nylon Connectors with PTFE Seals are threaded connectors used for coupling two items with a GPI finish.

  • Connectors have PTFE seals and FKM O-Rings
  • Blue glass-filled nylon
  • For use to 150°C
TRUSTED_SUSTAINABILITY

Specifications

Includes PTFE seals
Material Nylon
Thread Size 18-400 to 20-400
Product Type Connector
For Use With (Application) To connect standard rotary evaporator traps to vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.