Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Made from high quality borosilicate 3.3 glass
- Designed with ground joints to ensure tight connections with other pieces of glassware
- Ideal for transferring powder reagents
- 60° angle stem
- Autoclavable
Warranty and Services
1 year limited manufacturer warranty
Specifications
Specifications
| Diameter (Metric)Top | 70 mm |
| Material | Borosilicate Glass |
| Height (Metric) | 12.7 cm |
| Angle | 60° |
| Autoclavable | Autoclavable |
| Color | Clear |
| Description | Plain stem |
| Graduated | No |
| For Use With (Application) | Transferring liquids |
| Joint Number | 14/23 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
