Learn More
STERIS™ Glass-Klenz Glassware Detergent
Catalog No. 17010473 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
1 gal.
5 gal.
30 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-473 1 gal.
17-010-474 5 gal.
17-010-475 30 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-010-473 Supplier STERIS™ Supplier No. 111408
Add to Cart
Add to Cart
Glass-Klenz Detergent is a free rinsing glassware detergent specially formulated for use in automated glassware washing applications for the removal of particulate.
Glass-Klenz Detergent is a free rinsing glassware detergent specially formulated for use in automated glassware washing applications for the removal of particulate
HOW GLASS-KLENZ DETERGENT WORKS
Glass-Klenz Detergent is a low-foaming, chelant-based, liquid detergent for cleaning laboratory glassware.
WHY GLASS-KLENZ DETERGENT?
  • Flexibility - Effective removal of laboratory residues in both soft and hard water
  • Efficacy - Effectively holds soils in suspension allowing to be completely removed without redeposition
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Safety - Mild pH for enhanced safety and substrate compatibility
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Physical Properties: pH (1%): 10.9, typical
Specifications
Product Type Detergent
Color-coded No
Form Clear Liquid
Scent Unscented
Sterility Non-sterile
Size 1 gal.
Volume (Metric) 3.78 L