Learn More
Starna Cells Inc GLASS MICRO-CELL 1MM
SDP
Supplier:  Starna Cells Inc 21G1
Encompass_Preferred
Starna Standard Rectangular Cells with PTFE stopper, Window material Optical Glass, Usable range: 334 to 2500nm, Path Length: 1mm, Ext Width: 12.5mm, Ext Length: 3.5mm, Ext Height: 55mm, Int Width: 10mm, Int Length: 1mm
Catalog No. NC9167857