Learn More
Hoefer™ Glass Plates for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units
SDP
Catalog No. 03500206
Click to view available options
Type:
Glass Plate
Low Fluorescence Glass Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
03-500-206 Low Fluorescence Glass Plate
03-500-204 Glass Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-500-206 Supplier Hoefer™ Supplier No. SE6102LF
Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units
  • For use with SE660, SE600X Chroma, SE600, SE640, SE410, and SE400 Vertical Electrophoresis Units
  • Regular (straight, without notch)
  • Supplied in packs of 2
Specifications
For Use With (Equipment) SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit
Type Low Fluorescence Glass Plate
Material Glass
Quantity 2/Pk.
Dimensions (L x W) 18 x 16 cm
Length (Metric) 18 cm
Width (Metric) 16 cm