Learn More
Eisco™ Glass Prism and Lens Set
SCI_ED
Catalog No. S24391 Shop All Eisco Products
Change view
Click to view available options
Quantity:
7 Pieces
Catalog No. Quantity
S24391 7 Pieces
1 options
Catalog No. S24391 Supplier Eisco™ Supplier No. PH0572AG
Add to Cart
Add to Cart
For use in Physics optics experiments.
  • Made from high-quality glass
  • Set in wooden case with latches and custom-cut foam
Specifications
Product Type Prism Set, 7-Piece Glass
Lens Material Glass
For Use With (Application) Demonstrating the reflecting, refracting, and scattering properties of light
Includes 3 in. equilateral triangle, 3.5 in. dia. semicircle, trapezoid with 1.3 in. and 1.6 in. legs, 3.5 in. double convex prism w/3.75 in. inner radius, 3.5 in. double convex prism w/3.75 in. outer radius, 2 x 1.5 x 0.5 in. triangular prism, 2 x 1 x 0.75 in. triangular prism and wooden case
Quantity 7 Pieces