Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Glass Reagent Bottles with Hollow Interchangeable Hexagonal Glass Stoppers
SCI_ED

Catalog No. S39289 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
2 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S39289 60 mL
S39288 30 mL
S39290 125 mL
S39291 250 mL
S39292 500 mL
S39293 1 L
S39294 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S39289 Supplier Eisco™ Supplier No. CH0162B
Add to Cart
Add to Cart

Includes

Clear glass bottle, interchangeable hollow hexagonal glass stopper

Clear glass reagent bottles with interchangeable hollow glass stoppers are well suited for storing liquid solutions in the laboratory. Glass bottles and glass stoppers are autoclavable.

  • Made of high quality borosilicate 3.3 glass
  • Narrow mouth with interchangeable hollow hexagonal glass stopper
  • Bottle and stopper are autoclavable

Warranty and Services

1 year limited manufacturer warranty

Specifications

Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Clear
Description Hollow stopper
Diameter (English) 1.75 in.
For Use With (Application) Solution Preparation, Solution Storage
Includes Interchangeable hexagonal glass hollow stopper
Material Borosilicate Glass
Length (Metric) 44.5mm
Quantity 1
Sterility Non-sterile
Weight (Metric) 90.71 g
Width (Metric) 44.5mm
Closure Color Clear
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Cylindrical
Height (Metric) 114.3 mm
Product Type Reagent Bottle
For Use With (Equipment) Liquids
Height (English) 4.5 in.
Chemical Label Unlabeled
Closure Material Glass
Closure Size 14/23 mm
Diameter (Metric) 44.5 mm
Closure Type Stopper
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.