Learn More
Eisco™ Glass Right Angle Refraction Prism
SCI_ED
Catalog No. S27415 Shop All Eisco Products
Change view
Click to view available options
Material:
Glass
Catalog No. Material
S27415 Glass
1 options
Catalog No. S27415 Supplier Eisco™ Supplier No. PH0573B
Add to Cart
Add to Cart
Glass right angle refraction prisms are a great tool for studying the light spectrum.
  • Right angle refraction prism made from high quality glass
  • Creates a rainbow with any standard light source (ex. LED from a cellphone)
  • Designed for physics and educational demonstrations or experiments
  • Dimensions: 0.4 x 3.25 x 3.25 in.
Specifications
Material Glass
For Use With (Application) Physics and Educational Demonstrations or Experiments
Product Type Glass Right Angle Refraction Prism
Shape Triangular
Dimensions (L x W x H) 0.4 x 3.25 x 3.25 in.