Learn More
United Scientific Supplies Glass Separatory Funnel
Shop All United Scientific Supplies, Inc Products
Description
- These pear-shaped borosilicate glass funnels include a PTFE stopcock and a plastic stopper.
- Feature a marking spot and large white block letters for easy identification of capacity, as per ASTM E1096.
- PTFE stopcocks are compliant with ASTM standard E675.
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Material | Borosilicate Glass 3.3 |
| Color | Clear |
| Quantity | 2 Pk. |
| Product Type | Separatory Funnel |