Learn More
United Scientific Supplies Glass Separatory Funnel
SDPSCI_ED
Catalog No. S82836 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S82836 500 mL
S82832 50 mL
S82833 100 mL
S82834 125 mL
S82835 250 mL
S82837 1000 mL
S82838 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. S82836 Supplier United Scientific Supplies Supplier No. SF149500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Glass Separatory Funnels are pear shaped for easy handling and use.
  • These pear-shaped borosilicate glass funnels include a PTFE stopcock and a plastic stopper.
  • Feature a marking spot and large white block letters for easy identification of capacity, as per ASTM E1096.
  • PTFE stopcocks are compliant with ASTM standard E675.
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass 3.3
Color Clear
Quantity 2 Pk.
Product Type Separatory Funnel