Learn More
United Scientific™ Glass Streak Plates
SDPSCI_ED
Catalog No. S01057 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Dimensions (L x W):
2 x 1 in. (5.08 x 2.5 cm)
2 x 2 in. (5.08 x 5.08 cm)
3 x 2 in. (7.62 x 5.08 cm)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S01057 3 x 2 in. (7.62 x 5.08 cm)
S67817 2 x 1 in. (5.08 x 2.5 cm)
S67818 2 x 2 in. (5.08 x 5.08 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S01057 Supplier United Scientific™ Supplier No. GLP3X2P
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for hardness testing of rocks and minerals.
  • Designed for use in hardness testing of rocks and minerals, these 1/4 in. thick glass streak plates are sold as packs of 10
Specifications
For Use With (Application) Used for hardness testing of rocks and minerals
Product Type Streak Plate
Dimensions (L x W) 3 x 2 in. (7.62 x 5.08 cm)