Learn More
DWK Life Sciences Wheaton™ Glass Test Jar, 60mL
SDP
Catalog No. 03084174 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Liner:
PTFE
Polyvinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Liner
03-084-174 Polyvinyl
03-084-175 PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-084-174 Supplier DWK Life Sciences Supplier No. W216650
Add to Cart
Add to Cart
Test jars made of Type III Soda Lime glass with attached white polypropylene caps. Caps are lined with either polyvinyl or PTFE.
  • Smooth walls allow for complete removal of contents
  • Ideal for soil sampling and environmental applications
  • With attached screw caps lined in PTFE or PV
TRUSTED_SUSTAINABILITY
Specifications
Bottle Surface Smooth Walls
Color Clear
Quantity 180/Cs.
Closure Color White
Mouth Wide
Product Type Testing Jar
Closure Liner Polyvinyl
Closure Material Polypropylene