Learn More
Eisco™ Glass Vacuum Adapter Receivers
SCI_ED
Catalog No. S88402 Shop All Eisco Products
Change view
Click to view available options
Standard Taper Joint No.:
14/23
19/26
24/29
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
S88402 24/29
S88400 14/23
S88401 19/26
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S88402 Supplier Eisco™ Supplier No. CH01020C
Add to Cart
Add to Cart
Eisco™ Glass Vacuum Adapter Receivers feature screw threads to provide additional safety in lab.
  • With screw threads
  • Made from high quality Borosilicate 3.3 Glass
  • Screw threads provide additional lab safety
  • Screw thread joints allow flasks to connect firmly with other glassware
  • Autoclavable
Specifications
For Use With (Application) Distillation set ups
Height (Metric) 13 cm
Length (Metric) 20 cm
Material Borosilicate Glass (3.3)
Product Type Receiver, Vacuum
Quantity 1
Standard Taper Joint No. 24/29
Width (Metric) 14 cm