Learn More
Eisco™ Glass Volumetric Flask with Polypropylene Stopper, Sure Trace Certification
Catalog No. 12183050 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
10 mL
2000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-183-050 10 mL
12-183-049 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-183-050 Supplier Eisco™ Supplier No. CH0445A02ST
Volumetric Flask, Class A, ASTM. Polypropylene Stopper, Single, White Graduation. Made of Borosilicate Glass.
  • Made of high quality, heavy duty, Borosilicate 3.3 glass. Designed for use in professional laboratories, but is ideal for classroom, garage and home use
  • Features a single white graduation mark. Fitted with a polypropylene stopper
  • Class A tolerance. Compliant with ASTM-E288 standards
  • microwaveable and autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures & thermal shocks
  • Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind
Specifications
Capacity (Metric) 10 mL
Material Borosilicate Glass (3.3)
Product Type Volumetric Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Beaded
Sterility Non-sterile
Class Class A
Bottom Shape Flat
Surface Treatment Uncoated