Learn More
MilliporeSigma™ Supelco™ Glass Wool
Often used in GC inlet liners to improve sample vaporization, and/or to keep non-volatile material from entering the column
Supplier: MilliporeSigma™ Supelco™ 20411
Description
- Made up of short glass fibres
- Non-flammable and non-carcinogenic
- Widely considered as a thermal insulation material in ships, automobiles, air conditioning ducts, and water supply pipes
- Suitable as noise absorbing material in concert halls, studios, and movie theatres
- Can be used in packed GC columns, solvent desorption tubes, thermal desorption tubes, and purge traps to retain adsorbent beds
Specifications
GC Accessories and Adsorbents | |
Packed GC Columns, Solvent Desorption Tubes, Thermal Desorption Tubes, and Purge Traps to Retain Adsorbent Beds | |
Glass Fiber | |
50 g |