Learn More
MilliporeSigma™ Supelco™ GlasSeal™ Capillary Column Connector, Borosilicate Glass
Catalog No. 111020867 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
12/Pk.
Catalog No. Packaging Quantity
11-102-0867 12/Pk.
1 options
Catalog No. 11-102-0867 Supplier MilliporeSigma™ Supelco™ Supplier No. 20479
Add to Cart
Add to Cart
Column connectors are useful for attaching a guard column/retention gap to an analytical column, or for repairing a broken column.
Column connectors are useful for attaching a guard column/retention gap to an analytical column, or for repairing a broken column.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Gas Chromatography (GC)
Packaging Quantity 12/Pk.
GlasSeal is a trademark of Sigma-Aldrich Co. LLC