Learn More
Labconco™ Glassware Caps for RapidVap™ Evaporation Systems
SDP
Catalog No. 1631732 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Polyethylene
Catalog No. Material
16-317-32 Polyethylene
1 options
Catalog No. 16-317-32 Supplier Labconco™ Supplier No. 7925500
Add to Cart
Add to Cart
Placing Glassware Caps over the sample tubes prevents contamination and prevents moisture from condensing in low boiling point solvents.
For use with RapidVap borosilicate glassware tubes. Placing Caps on Tubes helps prevent cross contamination and moisture from condensing when using low boiling point solvents. The use of caps increases overall evaporation time 10 to 80 percent. Caps are available in polyethylene or PTFE. Eight polyethylene glassware caps are included with each RapidVap N2 Evaporation System.
Specifications
Material Polyethylene
Type Glassware Cap
For Use With (Equipment) 600mL glass tube of RapidVap™ evaporation system
Volume (Metric) 600 mL
Weight (English) 0.5 lb.
Weight (Metric) 0.2 kg