Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glassy carbon spherical powder, 200-400 micron, type 2

Catalog No. AA4255030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4255030 250 g
AA4255009 10 g
AA4255018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4255030 Supplier Thermo Scientific Chemicals Supplier No. 04255030
Add to Cart

Used as such in purifying metals, in electrodes, electrical devices & steel. Glassy Carbon Spherical Powder has numerous applications due its fundamental softness, including in lubricant formulations and as an additive to structural materials and metallic alloys.

Glassy carbon spherical powder is used as such in purifying metals, in electrodes, electrical devices & steel. Glassy Carbon Spherical Powder has numerous applications due its fundamental softness, including in lubricant formulations and as an additive to structural materials and metallic alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00133992

Specifications

Packaging Glass bottle
Quantity 250 g
UN Number UN1325
Solubility Information Insoluble in water.
Chemical Name or Material Glassy carbon spherical powder
What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.

Hazard Category H228
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Hazard Class: 4.1; Packaging Group: III
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.