Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glassy carbon splinter powder, 80-200 micron, type 1

Catalog No. AA3800230
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3800230 250 g
AA3800209 10 g
AA3800218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3800230 Supplier Thermo Scientific Chemicals Supplier No. 3800230

Glassy carbon splinter powder is used as such in purifying metals, in electrodes, electrical devices & steel. L-Cysteine methyl ester modified glassy carbon powder (CysOMe-GC) has been shown to have potential for the removal of toxic heavy metal ions, such as copper(II), cadmium(II) and arsenic(III), from real water samples. Amorphous carbon may be used to develop zero temperature coefficient of resistance (TCR) based systems. Glassy carbon has been used as a catalyst support for bimetallic Rh-ReOx. It may be used to enhance the accuracy of infrared (IR) surface temperature measurements on micro-electro-mechanical systems (MEMS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00133992

Specifications

Packaging Glass bottle
Quantity 250 g
UN Number UN1325
Solubility Information Insoluble in water.
Chemical Name or Material Glassy carbon splinter powder
EINECSNumber 231-153-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.