Learn More
Thermo Scientific™ Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Catalog No. 03251703 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Flowmeter Re-Calibration
Small Probe Adapter
Soft-sided Carry Case
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
03-251-703 Soft-sided Carry Case
03-251-704 Small Probe Adapter
14754509 Flowmeter Re-Calibration
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-251-703 Supplier Thermo Scientific™ Supplier No. 66002002
Add to Cart
Add to Cart
Probe adaptor and carrying case for use with the Thermo Scientific™ GLD Pro Gas Leak Detector.
Probe adaptor and carrying case for use with the Thermo Scientific™ GLD Pro Gas Leak Detector GLD Pro.
Soft-sided carrying case also for use with GFM Pro Handheld Equipment.
TRUSTED_SUSTAINABILITY
Specifications
Type Soft-sided Carry Case
For Use With (Equipment) GLD Pro Gas Leak Detector and GFM Pro Electric Flowmeter
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.