Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLEPP-1 Monoclonal Antibody (5F3F8), Abbomax

Catalog No. 502204196
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4196 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4196 Supplier Abbomax Inc Supplier No. 603900
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Kidney Cellular location: Membrane.

GLEPP-1 Monoclonal antibody specifically detects GLEPP-1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen GLEPP-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5F3F8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene PTPRO
Gene Accession No. Q16827
Gene Alias D28; FMI; GLEPP1; Glomerular epithelial protein 1; hPTP-J; NPHS6; osteoclastic transmembrane protein-tyrosine phosphatase; PCP2; PCP-2; phosphotyrosine phosphatase U2; protein tyrosine phosphatase PTP-U2; protein tyrosine phosphatase U2; protein tyrosine phosphatase, non-receptor type 15; protein tyrosine phosphatase, receptor type O; protein tyrosine phosphatase, receptor type, O; PTP phi; PTP pi; PTPase U2; PTPBK; PTP-J; Ptpn15; PTPoc; PTP-OC; PTPphi; Ptpro; PTP-RO; PTPROT; PTPRU; PTPU2; PTP-U2; receptor protein tyrosine phosphatase RO; receptor-type protein tyrosine phosphatase D30; receptor-type tyrosine-protein phosphatase O; RPTP-BK; R-PTP-O; R-PTP-psi; R-PTP-U
Host Species Mouse
Immunogen The recombinant fusion protein made from part of the extracellular domain of human GLEPP-1
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5800
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.