Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLG1 Mouse anti-Human, Clone: GLG1/970, Abnova™

Catalog No. 89034355 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-355 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-355 Supplier Abnova Corporation Supplier No. MAB14273
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native human GLG1.

  • High specificity
  • High reactivity

Specifications

Antigen GLG1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone GLG1/970
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human GLG1.
Dilution Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunocytochemistry (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The op
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. Q92896
Gene Alias CFR-1/ESL-1/FLJ23319/FLJ23967/MG-160/MG160
Gene Symbols GLG1
Host Species Mouse
Immunogen Golgi fraction from human liver cells.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2734
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.