Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ GLI-1 Antibody Blocking Peptide
SDP

Catalog No. NB398527 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398527 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398527 Supplier Novus Biologicals™ Supplier No. NBP178259PEP
Add to Cart
Add to Cart

Blocking Peptide

Specifications

Gene ID (Entrez) 2735
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
For Use With (Application) This peptide is useful as a blocking peptide for NBP1-78259. For further blocking peptide related protocol, click here .
Gene Alias GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI
Gene Symbol GLI1
Quantity 0.1 mg
Research Category Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators
Storage Buffer Peptide dissolved in dH20. Contains no BSA.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.