Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ GLI-2 Antibody Blocking Peptide
SDP

Catalog No. NB398468
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398468 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398468 Supplier Novus Biologicals™ Supplier No. NBP223602PEP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Blocking Peptide

Specifications

Gene ID (Entrez) 2736
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
For Use With (Application) This peptide is useful as a blocking peptide for NBP2-23602. For further blocking peptide related protocol, click here .
Gene Alias GLI family zinc finger 2, GLI-Kruppel family member GLI2, glioma-associated oncogene family zinc finger 2, HPE9tax helper protein 1, oncogene GLI2, Tax helper protein, tax helper protein 2, tax-responsive element-2 holding protein, tax-responsive element-25-bp sequence binding protein, THP, THP1, THP2, zinc finger protein GLI2
Gene Symbol GLI2
Quantity 0.1 mg
Research Category Alzheimers Research, Cancer, Cell Cycle and Replication, Neuroscience, Stem Cell Signaling Pathway
Storage Buffer Peptide dissolved in dH20. Contains no BSA.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.