Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLI3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528029 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528029 Supplier Invitrogen™ Supplier No. PA528029
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, mouse ESC, PC-12, Rat2. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (87%), Chicken (91%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein which belongs to the C2H2-type zinc finger proteins subclass of the Gli family. They are characterized as DNA-binding transcription factors and are mediators of Sonic hedgehog (Shh) signaling. The protein encoded by this gene localizes in the cytoplasm and activates patched Drosophila homolog (PTCH) gene expression. It is also thought to play a role during embryogenesis. Mutations in this gene have been associated with several diseases, including Greig cephalopolysyndactyly syndrome, Pallister-Hall syndrome, preaxial polydactyly type IV, and postaxial polydactyly types A1 and B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLI3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene GLI3
Gene Accession No. P10071, Q61602
Gene Alias ACLS; add; AI854843; anterior digit pattern deformity; AU023367; Bph; brachyphalangy; extra toes; GCPS; GLI family zinc finger 3; GLI family zinc finger 3 L homeolog; gli3; GLI3 C-terminally truncated form; GLI3 form of 190 kDa; GLI3 form of 83 kDa; GLI3 full length protein; GLI3 full-length protein; gli3.L; GLI3-190; GLI3-83; GLI3FL; GLI-Kruppel family member 3; GLI-Kruppel family member GLI3; GLI-Kruppel family member GLI3 (Greig cephalopolysyndactyly syndrome); glioma-associated oncogene family zinc finger 3; im:7142603; neural specific DNA binding protein; neural-specific DNA-binding protein xGLI3; oncogene GLI3; PAPA; PAP-A; PAPA1; PAPB; Pdn; PHS; polydactyly Nagoya; PPDIV; Transcriptional activator GLI3; Transcriptional repressor GLI3R; XELAEV_18031191mg; xgli3; xGLI-3; Xt; Zinc finger protein GLI3; zinc finger protein GLI3; transcriptional activator GLI3; zinc finger transcription factor Gli3
Gene Symbols GLI3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GLI3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140588, 14634, 2737
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.