Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gliadin Recombinant Mouse Monoclonal Antibody (4H12)

Catalog No. MA566472
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA566472 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA566472 Supplier Invitrogen™ Supplier No. MA566472
Only null left
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Gliadin Recombinant Monoclonal Antibody for ELISA

Celiac disease is associated with a CD4+ T-cell response to epitopes of gliadin presented by HLA-DQ2 or -DQ8 class II MHC molecules. These epitopes are present in a 33-mer peptide of wheat alpha-gliadin, residues 56-88, which is resistant to digestion and forms a substrate for tissue transglutaminase (TG2), generating the glutamic acid residues essential for binding to HLA-DQ2. Complete homology exists between residues 63-73, 70-80 and 77-87 of wheat alpha gliadin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gliadin
Applications ELISA
Classification Recombinant Monoclonal
Clone 4H12
Concentration 10.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% NeoCide PC-950
Gene Alias AGP; gliadin peptide
Host Species Mouse
Immunogen Gliadin.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.