Learn More
CPC Scientific H-Tyr-Pro-Gln-Pro-Gln-Pro-Phe-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific OPIO012A
Encompass
Gliadorphin-7 is an opioid peptide which is formed during digestion of the gliadin component of the gluten protein. Elevated concentrations of gliadinoorphin-7 due to insufficient proteolysis has been associated with autism, schizophrenia, and celiac disease.ONE-LETTER SEQUENCE: YPQPQPFMOLECULAR FORMULA: C43H57N9O11MOLECULAR WEIGHT:875.98STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [107936-65-2], RESEARCH AREA: PainREFERENCES: D.G.Payan et al., Life Sci., 40, 1229 (1987); L.Graf et al., Neuropeptides, 9, 113 (1987); M.Hollosi et al., Neuropeptides, 17, 111 (1990); Z.Sun and R.Cade, Peptides, 24, 321 (2003)
Catalog No. 50-210-9986
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.