Learn More
Creative Biomart Glial Growth Factor (GGF2) Protein
Supplier:  Creative Biomart THP0237
Encompass_Preferred
Recombinant human GGF2 (Glial growth factor) is a disulfide-linked monomeric protein consisting of 315 amino acid residues, and migrates as an approximately 34 kDa protein under non-reducing and reducing conditions in SDS-PAGE. Optimized DNA sequence encoding Human Neuregulin-1 isoform 9 (GGF2) was expressed in E. coli.
Catalog No. 50-208-4288
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.