Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glibenclamide 98.5+%, TCI America™
SDP

Catalog No. G038225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G038225G 25 g
G03825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G038225G Supplier TCI America Supplier No. G038225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10238-21-8
Molecular Formula C23H28ClN3O5S
Molecular Weight (g/mol) 494.003
MDL Number MFCD00056625
InChI Key ZNNLBTZKUZBEKO-UHFFFAOYSA-N
Synonym glyburide, glibenclamide, glybenclamide, micronase, diabeta, glynase, daonil, euglucon, maninil, semi-daonil
PubChem CID 3488
ChEBI CHEBI:5441
IUPAC Name 5-chloro-N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-2-methoxybenzamide
SMILES COC1=C(C=C(C=C1)Cl)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 25 g
Formula Weight 494.00
Percent Purity ≥98.5% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Glibenclamide
EINECSNumber (9)-0412
RTECSNumber YS4725200
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.