Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Global RNA Methylation Assay Kit, 5-Methyl Cytosine, Fluorometric, 1x96 Tests

Catalog No. 502586669
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-6669 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-6669 Supplier Abcam Supplier No. AB2334921X96TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Fluorometric assay kit designed for quantitative analysis of global RNA 5-methylcytosine methylation.

  • Detection Method: Fluorescent
  • Results Type: Quantitative
  • Usage: Suitable for use with microplate reader
  • Citations: Cited in 1 publication

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule 5-Methylcytosine (RNA)
No. of Reactions 96 Tests
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Format Kit
Quantity 96 Tests
For Use With (Application) RNA methylation analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.