Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Global RNA Methylation Assay Kit, Fluorometric, 48 Tests

Catalog No. 502586670
Encompass_Preferred
Change view
Click to view available options
Quantity:
48 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-6670 48 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-6670 Supplier Abcam Supplier No. AB2334921X48TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorometric assay kit designed for the quantitative detection of global RNA methylation (5-methylcytosine) using a microplate reader.

  • Detection Method: Fluorescent
  • Assay Type: Quantitative
  • Target: 5-methylcytosine (5-mC) RNA methylation
  • Compatible Instrumentation: Microplate reader
  • Citation Count: 1

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule 5-Methylcytosine (RNA)
For Use With (Equipment) Microplate Reader
Format Kit
No. of Reactions 48 Tests
Detection Method Fluorescence
Quantity 48 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.