Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GlobalFiler™ Express PCR Amplification Kit

Catalog No. 501607771
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
200 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
50-160-7771 200 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-160-7771 Supplier LIFE TECHNOLOGIES Supplier No. 4476609
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Specifications

Content And Storage Store at -15°C to -25°C upon receipt and 2 to 8°C after initial use for up to 6 months.
Product Line GlobalFiler
Product Type PCR Amplification Kit
Quantity 200 reactions
Shipping Condition Dry Ice
No. of Reactions 200 Reactions
Starting Material DNA
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.