Learn More
Globe Scientific Dispenser For Borosilicate Glass Culture Tubes; for 75mm Tubes
Catalog No. 21200206 Shop All Globe Scientific Products
Change view
Click to view available options
Capacity (Metric):
For 10 x 75mm and 12 x 75mm tubes
Catalog No. Capacity (Metric)
21-200-206 For 10 x 75mm and 12 x 75mm tubes
1 options
Catalog No. 21-200-206 Supplier Globe Scientific Supplier No. 1590
Add to Cart
Add to Cart
Metal Dispenser, for 10 x 75mm and 12 x 75mm tubes
Metal dispenser
Heavy Duty dispenser
  • Made from white painted metal and feature a flexi-glass cover that pulls up to dispense the tubes
  • Each dispenser holds one box of 250 glass tubes
Specifications
Certifications/Compliance ISO 9001:2008, ASTM E 890, Type 1
Capacity (Metric) For 10 x 75mm and 12 x 75mm tubes
Material Metal