Learn More
Globe Scientific Pediatric Micro Sample Cup
Catalog No. 21200249 Shop All Globe Scientific Products
Change view
Click to view available options
Material:
Polystyrene
Catalog No. Material
21-200-249 Polystyrene
1 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. 21-200-249 Supplier Globe Scientific Supplier No. 110915
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
For use on automated instrumentation for Roche™ analyzers
  • Made of polystyrene
  • Also for use on Hitachi, Elecsys, Cobas Integra and Core analyzers
  • Dimensions: 0.6 x 1.45 in. (17 x 37mm)
Specifications
Product Type Sample Cup
Material Polystyrene
For Use With (Equipment) Automated Roche™, Hitachi™, Elecsys™, Cobas Integra™ and Core™ analyzers