Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Sample Cups for Ciba Corning™ 550 Express

Catalog No. 21200226 Shop All Globe Scientific Products
Change view
Click to view available options
Capacity (Metric):
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
21-200-226 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-226 Supplier Globe Scientific Supplier No. 5520

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use on automated instrumentation including 550 Express, Express Plus analyzers

Specifications

Capacity (Metric) 1 mL
Product Type Sample Cap
Capacity (English) 0.03 oz.
Material Polystyrene
For Use With (Equipment) Automated instrumentation including 550 Express, express plus analyzers
Capacity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.