Learn More
Poltex Glove Box Holder with Bin
Description
- Rubber bumpons on base for non-slip placement on countertop.
- Choose from wall mount/countertop placement (screws/anchors included), magnet mount (powerful magnets included), or tape mount (VHB tape included).
Specifications
Specifications
| Product Type | Glove Box Holder |
| Color | Clear |
| Material | PETG |
| Mounting | VHB Tape |
| Capacity | 2 Boxes |