Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Clear Acrylic Glove Box Dispenser

Catalog No. 19129469 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity:
1 Box
2 Boxes
3 Boxes
Dimensions (L x W x H):
10.4 x 4.1 x 10 in.
10.4 x 4.1 x 15 in.
10.9 x 4.1 x 5 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Dimensions (L x W x H)
19-129-469 2 Boxes 10.4 x 4.1 x 10 in.
19-129-468 1 Box 10.9 x 4.1 x 5 in.
19-129-470 3 Boxes 10.4 x 4.1 x 15 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-129-469 Supplier Brady™ Supplier No. GD02
Add to Cart
Add to Cart
This item is not returnable. View return policy

Remind employees to wear their gloves by mounting glove dispensers near all hazardous work areas.

  • Dispensers are constructed of clear acrylic for stock visibility
  • Designed to be the perfect size for standard glove boxes
  • Can be mounted vertically on a wall or counter
  • Gloves boxes are not included with dispenser purchase

Specifications

Product Type Glove Box Holder
Color Clear
Dimensions (L x W x H) 10.4 x 4.1 x 10 in.
Holds Disposable Glove Boxes
Material Acrylic
No. of Compartments 1
Product Style Top Loading
Capacity 2 Boxes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.