Learn More
Med Vet International Exam Glove McKesson Confiderm LDC Medium NonSterile Nitrile Standard Cuff Length Fully Textured Black Chemo Tested / Fentanyl Tested, 14-6N54C, 2500/Case
Supplier:  Med Vet International 1159334CS
Encompass_Preferred
Low dermatitis potential; ideal for individuals with skin sensitivities to regular nitrile gloves Tested to resist permeation of 15 chemotherapy drugs, plus fentanyl Exterior's fully textured finish promotes slip resistance in wet or dry conditions Thin with high-tactile sensitivity Excellent conformability provides superior fit and extended user comfort Durability and tensile strength thanks to nitrile construction Fingertip thickness: 0.07mm (2.8 mil) Chlorinated internal surface Medium
Catalog No. 50-304-3213
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.