Learn More
Glove Guard™ Glove Clip
Catalog No. 19811715
Change view
Click to view available options
Color:
Hi-Viz Yellow
Specify when ordering
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-811-715 Hi-Viz Yellow
19-811-709 Specify when ordering
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-811-715 Supplier Glove Guard™ Supplier No. 1939HVYWBP
Attaches to a belt loop or directly to clothing
  • Keeps work gloves with employees to help reduce recordable hand injuries
  • Secure locking provides resilience and “creep” resistance
  • Resists fatigue, mechanical damage, and chemical attack
  • Completely dielectric
  • Patented safety breakaway
  • Complements corporate PPE policies and Voluntary Protection Programs
Order Info
Available in yellow, blue, high-vis yellow, granite, red, olive green, black, green, lime green and blaze orange; specify when ordering.
Specifications
Color Hi-Viz Yellow