Learn More
Med Vet International SW PowerForm PF-95BL Blue X-large Powder Free Disposable Gloves - Medical Exam Grade - 9.5 in Length - Textured Fingers Finish - 5.0 mil Thick - N700345, 100/Box
Supplier:  Med Vet International N700345
Encompass_Preferred
Product SpecificationsBrand: SW SafetyProduct Type: Disposable GlovesGrade: Medical ExamMaterial: NitrilePowdering: Powder FreeThickness: 5.0 milOverall Length: 9.5 inColor: BlueSize: X-LargeSurface Finish: Textured FingersResistant Properties: Chemical-ResistantCuff Type: BeadedSeries: PF-95BLStandards Met: SHA Dermatologically ApprovedPackage Quantity: 100 per box, 10 boxes per caseTrade Name: PowerForm, Dry Core• Dry Core absorbent technology keeps hands dry and cool• Fully textured surface delivers added grip• Beaded cuff allows easy donning and doffing• 100% air inspected / consistent thickness tip to cuff• ANSI Level 3 abrasion resistance
Catalog No. 50-252-2227
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.