Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8365 Type R Class 3 High Visibility Rain Jacket

Catalog No. 19820822 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-820-822 Orange Small
19-820-824 Orange Medium
19-820-825 Orange Large
19-820-826 Orange X-Large
19-820-827 Orange 2X-Large
19-820-840 Orange 3X-Large
19-820-841 Orange 4X-Large
19-820-842 Orange 5X-Large
19-820-843 Lime Small
19-820-844 Lime Medium
19-820-845 Lime Large
19-820-846 Lime X-Large
19-820-847 Lime 2X-Large
19-820-848 Lime 3X-Large
19-820-849 Lime 4X-Large
19-820-851 Lime 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-820-822 Supplier Ergodyne™ Supplier No. 24312
Add to Cart
Add to Cart
This item is not returnable. View return policy

Class 3 Rain Jacket constructed of a breathable poly shell designed to keep the rain out while letting perspiration escape.

  • Weatherproof 300D oxford polyester outer shell
  • Breathable PU coating with polyester mesh lining
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Inset hood with drawstring and stoppers
  • Two external front pockets with snap flaps
  • Chest pocket
  • Dual mic tabs for clipping radios
  • YKK #5 zippers

Specifications

Color Orange
Material Oxford Polyester (300D)
Size Small
No. of Pockets 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.