Learn More
Description
- Weatherproof 300D oxford polyester outer shell
- Breathable PU coating with polyester mesh lining
- 2 in ANSI-compliant 3M™ Scotchlite™ Reflective
- Black panels hide dirt and wear
- Inset hood with drawstring and stoppers
- Two external front pockets with snap flaps
- Cell phone chest pocket
- Dual mic tabs for clipping radios
- YKK #5 zippers
Specifications
Specifications
| Size | 2X-Large |
| Material | Oxford Polyester (300D) |
| Color | Lime |
| For Use With (Application) | Flagging Crew, Road Construction, Utility Crew, Emergency Response |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.