Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8377 Type R Class 3 Quilted Thermal High Visibility Jacket

Catalog No. 19910138 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-910-138 Orange 2X-Large
19-910-134 Orange Small
19-910-135 Orange Medium
19-910-136 Orange Large
19-910-137 Orange X-Large
19-910-139 Orange 3X-Large
19-910-140 Orange 4X-Large
19-910-141 Orange 5X-Large
19-910-142 Lime Small
19-910-143 Lime Medium
19-910-144 Lime Large
19-910-145 Lime X-Large
19-910-146 Lime 2X-Large
19-910-147 Lime 3X-Large
19-910-148 Lime 4X-Large
19-910-149 Lime 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-910-138 Supplier Ergodyne™ Supplier No. 25616
Add to Cart
Add to Cart
This item is not returnable. View return policy

GloWear™ 8377 Type R Class 3 Quilted Thermal High Visibility Jacket features a polyester outer shell with PU coating that resists water and wind while staying breathable for all day comfort.

  • Wind and water resistant
  • Body and sleeves are lined with 160 g thermal quilted insulation for warmth without the extra bulk
  • 2 in. ANSI-compliant reflective material for night-time conspicuity
  • Cell phone chest pocket, dual slot pen pocket on arm, dual slash pockets and dual mic tabs for clipping radios
  • Black panels on the sleeves and front of jacket to hide dirt and wear
  • Inset hood with drawstring and stoppers
  • Machine washable

Specifications

Size 2X-Large
Material Oxford Polyester (300D)
Color Orange
Coating Polyurethane
Liner Material 160 g thermal quilted insulation
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.