Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8379 Type R Class 3 Fleece-Lined High Visibility Bomber Jacket

Catalog No. 19820896 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-820-896 Lime 2X-Large
19-820-884 Orange Small
19-820-885 Orange Medium
19-820-886 Orange Large
19-820-887 Orange X-Large
19-820-888 Orange 2X-Large
19-820-889 Orange 3X-Large
19-820-890 Orange 4X-Large
19-820-891 Orange 5X-Large
19-820-892 Lime Small
19-820-893 Lime Medium
19-820-894 Lime Large
19-820-895 Lime X-Large
19-820-899 Lime 5X-Large
19-820-897 Lime 3X-Large
19-820-898 Lime 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-820-896 Supplier Ergodyne™ Supplier No. 24476
This item is not returnable. View return policy

GloWear™ 8379 Type R Class 3 Fleece-Lined High Visibility Bomber Jacket has a zip-out fleece liner and can be worn three different ways for ultimate adaptability.

  • Wind and water resistant
  • Zip-out 220 g microfleece liner can be worn on its own or zipped into the shell for warmth without the extra bulk
  • 2 in. ANSI-compliant reflective material for night-time conspicuity
  • Cell phone chest pocket, dual slot pen pocket on arm, dual slash pockets and dual mic tabs for clipping radios
  • Black panels on the sleeves and front of jacket to hide dirt and wear
  • Inset hood with drawstring and stoppers
  • Machine washable

Specifications

Size 2X-Large
Material Oxford Polyester (300D)
Color Lime
Coating Polyurethane
Liner Material 220 g microfleece
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.