Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8381 3-in-1, Type R, Class 3, Thermal High Visibility Bomber Jacket

Catalog No. 19910125 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-910-125 Orange 5X-Large
19-910-118 Orange Small
19-910-119 Orange Medium
19-910-120 Orange Large
19-910-121 Orange X-Large
19-910-122 Orange 2X-Large
19-910-123 Orange 3X-Large
19-910-124 Orange 4X-Large
19-910-126 Lime Small
19-910-127 Lime Medium
19-910-128 Lime Large
19-910-129 Lime X-Large
19-910-130 Lime 2X-Large
19-910-131 Lime 3X-Large
19-910-132 Lime 4X-Large
19-910-133 Lime 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-910-125 Supplier Ergodyne™ Supplier No. 25589
Add to Cart
Add to Cart
This item is not returnable. View return policy

GloWear™ 8381 Type R Class 3, 3-in-1 High Visibility Bomber Jacket can be worn in a variety of ways for ultimate adaptability in changing weather conditions.

  • 300D performance ripstop polyester outer shell with PU coating resists water and wind while still being breathable for comfort
  • 280 g fleece body and thermal insulated, quilted sleeves for warmth without the extra bulk
  • Zip-out fleece liner can be worn on its own, or zip-off the sleeves to create a fleece vest
  • 2 in. ANSI-compliant reflective material for night-time conspicuity
  • Black panels on sleeves hide dirt and wear
  • Inset hood with drawstring and stoppers can be removed
  • Large hidden external zipper pocket for secure storage
  • External zipper chest pocket, dual slot pen pocket on arm, and dual mic tabs for clipping radios
  • Storm flap snap front covers front zipper
  • Machine washable

Specifications

Size 5X-Large
Material Ripstop Polyester (300D)
Color Orange
Coating Polyurethane
Liner Material 280 g thermal-insulated fleece body
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.