Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8384 Type R Class 3 Thermal Quilted High Visibility Jacket

Catalog No. 19910103 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-910-103 Lime Medium
19-910-102 Lime Small
19-910-104 Lime Large
19-910-109 Lime 5X-Large
19-910-110 Orange Small
19-910-111 Orange Medium
19-910-112 Orange Large
19-910-113 Orange X-Large
19-910-114 Orange 2X-Large
19-910-115 Orange 3X-Large
19-910-116 Orange 4X-Large
19-910-117 Orange 5X-Large
19-910-105 Lime X-Large
19-910-106 Lime 2X-Large
19-910-107 Lime 3X-Large
19-910-108 Lime 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-910-103 Supplier Ergodyne™ Supplier No. 25563
Add to Cart
Add to Cart
This item is not returnable. View return policy

GloWear™ 8384 Type R Class 3 Thermal Quilted High Visibility Jacket is an ideal warm winter work jacket designed for safety on the job.

  • 300D oxford outer shell with PU coating resists wind and water to keep you dry
  • Body and sleeves are lined with 160 g thermal quilted insulation for warmth without the extra bulk
  • 2 in. ANSI-compliant reflective material for night-time conspicuity
  • Internal waist cord cinches tight to keep out the cold
  • Cell phone chest pocket, dual-slot pen pocket on arm, and dual-mic tabs for clipping radios
  • Black panels on the sleeves and front of the jacket to hide dirt and wear
  • Inset hood with drawstring and stoppers
  • Machine washable

Specifications

Size Medium
Material Oxford Polyester (300D)
Color Lime
Coating Polyurethane
Liner Material 160 g thermal quilted insulation
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.