Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8386 Type R Class 3 High Visibility Outer Shell Jacket

Catalog No. 19820954 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-820-954 Orange 3X-Large
19-820-929 Lime Small
19-820-930 Lime Medium
19-820-931 Lime Large
19-820-932 Lime X-Large
19-820-934 Lime 2X-Large
19-820-936 Lime 3X-Large
19-820-938 Lime 4X-Large
19-820-939 Lime 5X-Large
19-820-949 Orange Small
19-820-950 Orange Medium
19-820-951 Orange Large
19-820-952 Orange X-Large
19-820-953 Orange -
19-820-955 Orange 4X-Large
19-820-956 Orange 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-820-954 Supplier Ergodyne™ Supplier No. 25467
Add to Cart
Add to Cart
This item is not returnable. View return policy

Class 3 Outer Shell Jacket is a breathable poly shell that keeps the rain out while letting perspiration escape.

  • Weatherproof 300D oxford polyester outer shell
  • Breathable PU coating with polyester mesh lining
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Black panels hide dirt and wear
  • Inset hood with drawstring and stoppers
  • Two external front pockets with snap flaps
  • External zipper chest pocket
  • Cellphone chest pocket
  • Clear ID badge holder
  • Dual mic tabs for clipping radios
  • YKK #5 zippers

Specifications

Size 3X-Large
Color Orange
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.